siRNA Details
Gene Symbol: LOC691909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134694.2 | NP_001128166.2 | 1 | 490 | ||
GenBank mRNA | BC158625.1 | - | 1 | 490 | 7454926 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC691909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134694.2 | NP_001128166.2 | 1 | 490 | ||
GenBank mRNA | BC158625.1 | - | 1 | 490 | 7454926 |