siRNA Details
| Gene Symbol: LOC691952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109652.1 | NP_001103122.1 | 6 | 468 | ||
| XM_008762201.1 | XP_008760423.1 | 6 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691952| Gene Symbol: LOC691952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109652.1 | NP_001103122.1 | 6 | 468 | ||
| XM_008762201.1 | XP_008760423.1 | 6 | 670 | |||