siRNA Details
| Gene Symbol: Ska2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009624.1 | NP_001009624.1 | 3 | 291 | ||
| GenBank mRNA | BC088264.1 | - | 3 | 291 | 7377141 | |
| FQ220730.1 | - | 3 | 290 | |||
| FQ229832.1 | - | 3 | 290 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ska2| Gene Symbol: Ska2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009624.1 | NP_001009624.1 | 3 | 291 | ||
| GenBank mRNA | BC088264.1 | - | 3 | 291 | 7377141 | |
| FQ220730.1 | - | 3 | 290 | |||
| FQ229832.1 | - | 3 | 290 | |||