siRNA Details
| Gene Symbol: Slc35g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127658.1 | NP_001121130.1 | 2 | 680 | ||
| GenBank mRNA | BC159402.1 | - | 2 | 680 | 7457005 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35g3| Gene Symbol: Slc35g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127658.1 | NP_001121130.1 | 2 | 680 | ||
| GenBank mRNA | BC159402.1 | - | 2 | 680 | 7457005 | |