siRNA Details
| Gene Symbol: LOC691995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103353.2 | NP_001096823.1 | 1 | 905 | ||
| GenBank mRNA | BC087739.1 | - | 1 | 873 | ||
| BC110041.1 | - | 1 | 884 | 7932837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691995| Gene Symbol: LOC691995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103353.2 | NP_001096823.1 | 1 | 905 | ||
| GenBank mRNA | BC087739.1 | - | 1 | 873 | ||
| BC110041.1 | - | 1 | 884 | 7932837 | ||