siRNA Details
| Gene Symbol: AMELX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142.2 | NP_001133.1 | 5 | 608 | ||
| NM_182680.1 | NP_872621.1 | 6 | 650 | |||
| NM_182681.1 | NP_872622.1 | 4 | 560 | |||
| XM_017029404.1 | XP_016884893.1 | 4 | 1709 | |||
| GenBank mRNA | AF436849.1 | - | 6 | 650 | ||
| BC069118.1 | - | 5 | 608 | 7216972 | ||
| BC074951.2 | - | 5 | 587 | 30915277 | ||
| M86932.1 | - | 5 | 608 | |||
| S67147.1 | - | 1 | 396 | |||

AMELX
Genomic Map