siRNA Details
| Gene Symbol: GAS8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122031.1 | 1 | 891 | |||
| GenBank mRNA | AF050080.1 | - | 1 | 332 | ||
| AF050081.1 | - | 1 | 914 | |||
| BC093751.1 | - | 1 | 357 | 7939596 | ||
| BC104849.1 | - | 1 | 357 | 8143852 | ||

GAS8-AS1
Genomic Map