siRNA Details
| Gene Symbol: CD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006137.6 | NP_006128.1 | 3, 4 | 719 | ||
| XM_011523608.1 | XP_011521910.1 | 3, 4 | 686 | |||
| XM_017025316.1 | XP_016880805.1 | 3, 4 | 752 | |||
| GenBank mRNA | AK223465.1 | - | 3, 4 | 691 | ||
| AY935534.1 | - | 3, 4 | 691 | |||
| AY935536.1 | - | 3, 4 | 691 | |||
| AY935537.1 | - | 3, 4 | 679 | |||
| BC009293.2 | - | 3, 4 | 663 | 4120480 | ||
| BC013297.2 | - | 3, 4 | 663 | 3529600 | ||
| BI838777.1 | - | 3, 4 | 721 | |||
| BT006696.1 | - | 3, 4 | 610 | |||
| X06180.1 | - | 2, 3 | 1097 | |||

CD7
Genomic Map