siRNA Details
| Gene Symbol: GNRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310220.1 | NP_001297149.1 | 2 | 144 | ||
| NM_001501.1 | NP_001492.1 | 2 | 149 | |||
| NM_178331.1 | NP_847901.1 | 2 | 149 | |||
| NM_178332.1 | NP_847902.1 | 2 | 149 | |||
| XM_017027823.1 | XP_016883312.1 | 2 | 2756 | |||
| GenBank mRNA | AF036330.1 | - | 2 | 149 | ||
| AF190640.1 | - | 1 | 20 | |||
| BC069362.1 | - | 2 | 148 | 7262673 | ||
| BC115399.1 | - | 2 | 148 | 40069240 | ||
| BC115400.1 | - | 2 | 143 | 40069241 | ||

GNRH2
Genomic Map