siRNA Details
| Gene Symbol: GP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000174.4 | NP_000165.1 | 3 | 849 | ||
| XM_005247374.3 | XP_005247431.1 | 5 | 1145 | |||
| XM_011512701.1 | XP_011511003.1 | 6 | 1227 | |||
| XM_011512702.1 | XP_011511004.1 | 5 | 1102 | |||
| GenBank mRNA | BC030229.1 | - | 3 | 814 | 5226377 | |
| M25827.1 | - | 1 | 588 | |||
| X52997.1 | - | 3 | 846 | |||

GP9
Genomic Map