siRNA Details
| Gene Symbol: GPR25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005298.3 | NP_005289.2 | 1 | 1113 | ||
| GenBank mRNA | AK315339.1 | - | 1 | 1087 | ||
| BC126386.1 | - | 1 | 1113 | 8992102 | ||
| BC126388.1 | - | 1 | 1113 | 8992104 | ||
More Information
Set Membership: |
|

GPR25
Genomic Map