siRNA Details
| Gene Symbol: GPR31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005299.2 | NP_005290.2 | 1 | 367 | ||
| GenBank mRNA | BC095537.1 | - | 1 | 364 | ||
| CD246167.1 | - | 1 | 375 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR31| Gene Symbol: GPR31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005299.2 | NP_005290.2 | 1 | 367 | ||
| GenBank mRNA | BC095537.1 | - | 1 | 364 | ||
| CD246167.1 | - | 1 | 375 | |||