siRNA Details
| Gene Symbol: FFAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005303.2 | NP_005294.1 | 1 | 815 | ||
| GenBank mRNA | BC095536.1 | - | 1 | 802 | ||
| BC120943.1 | - | 1 | 802 | |||
| BC120944.1 | - | 1 | 805 | 40115111 | ||
| EF611308.1 | - | 1 | 805 | |||

FFAR1
Genomic Map