siRNA Details
| Gene Symbol: IGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000597.2 | NP_000588.2 | 4 | 955 | ||
| NM_001313990.1 | NP_001300919.1 | 4 | 513 | |||
| NM_001313992.1 | NP_001300921.1 | 4 | 832 | |||
| NM_001313993.1 | NP_001300922.1 | 4 | 571 | |||
| GenBank mRNA | A09809.1 | - | 4 | 954 | ||
| AB209509.1 | - | 2 | 3941 | |||
| BC004312.1 | - | 4 | 929 | 3627826 | ||
| BC009902.1 | - | 4 | 931 | 3509749 | ||
| BC012769.1 | - | 4 | 929 | 3689990 | ||
| BC071967.1 | - | 4 | 942 | 6650239 | ||
| M35410.1 | - | 4 | 898 | |||
| X16302.1 | - | 4 | 952 | |||

IGFBP2
Genomic Map