siRNA Details
| Gene Symbol: LY6E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127213.1 | NP_001120685.1 | 4 | 445 | ||
| NM_002346.2 | NP_002337.1 | 4 | 450 | |||
| GenBank mRNA | AF085875.1 | - | 2 | 186 | ||
| AK129615.1 | - | 4 | 407 | |||
| AK223074.1 | - | 4 | 429 | |||
| AK311983.1 | - | 4 | 406 | |||
| BC003392.1 | - | 4 | 354 | |||
| BC119708.1 | - | 3 | 331 | 40117169 | ||
| BC119709.1 | - | 3 | 331 | 40117170 | ||
| DB458093.1 | - | 4 | 450 | |||
| U42376.1 | - | 4 | 385 | |||
| U56145.1 | - | 3 | 291 | |||
| Z68179.1 | - | 4 | 385 | |||

LY6E
Genomic Map