siRNA Details
| Gene Symbol: OPN1SW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001708.2 | NP_001699.1 | 3 | 590 | ||
| GenBank mRNA | DQ822478.1 | - | 3 | 590 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OPN1SW| Gene Symbol: OPN1SW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001708.2 | NP_001699.1 | 3 | 590 | ||
| GenBank mRNA | DQ822478.1 | - | 3 | 590 | ||
Set Membership: |
|