siRNA Details
| Gene Symbol: MT1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301267.1 | NP_001288196.1 | 3 | 285 | ||
| NM_005950.2 | NP_005941.1 | 3 | 282 | |||
| GenBank mRNA | BC020757.1 | - | 3 | 285 | 4767886 | |
| BC048334.1 | - | 1 | 1198 | |||
| BM817643.1 | - | 3 | 285 | |||
| CD175975.1 | - | 3 | 301 | |||

MT1G
Genomic Map