siRNA Details
Gene Symbol: NEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005383.2 | NP_005374.2 | 2 | 880 | ||
GenBank mRNA | BC069151.1 | - | 2 | 880 | 7217011 | |
BC107053.2 | - | 2 | 880 | 40004620 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005383.2 | NP_005374.2 | 2 | 880 | ||
GenBank mRNA | BC069151.1 | - | 2 | 880 | 7217011 | |
BC107053.2 | - | 2 | 880 | 40004620 |