siRNA Details
| Gene Symbol: POLR2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021128.4 | NP_066951.1 | 2 | 631 | ||
| GenBank mRNA | BC018649.2 | - | 2 | 612 | 2958832 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR2L| Gene Symbol: POLR2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021128.4 | NP_066951.1 | 2 | 631 | ||
| GenBank mRNA | BC018649.2 | - | 2 | 612 | 2958832 | |