siRNA Details
| Gene Symbol: PRSS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197097.2 | NP_001184026.2 | 4 | 633 | ||
| NM_001197098.1 | NP_001184027.1 | 3 | 275 | |||
| NM_002771.3 | NP_002762.2 | 3 | 296 | |||
| NM_007343.3 | NP_031369.2 | 3 | 505 | |||
| XM_011517965.1 | XP_011516267.1 | 4 | 556 | |||
| GenBank mRNA | AB298285.1 | - | 3 | 505 | ||
| AB298286.1 | - | 3 | 275 | |||
| AY052783.1 | - | 4 | 410 | |||
| BC030238.1 | - | 3 | 295 | |||
| BC069476.1 | - | 3 | 296 | 7262223 | ||
| BC069494.1 | - | 3 | 296 | 7262235 | ||
| D45417.1 | - | 3 | 283 | |||
| X15505.1 | - | 3 | 296 | |||
| X71345.1 | - | 3 | 323 | |||
| X72781.1 | - | 3 | 326 | |||

PRSS3
Genomic Map