siRNA Details
| Gene Symbol: PTPRCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005608.2 | NP_005599.1 | 2 | 163 | ||
| GenBank mRNA | AK312022.1 | - | 2 | 163 | ||
| BC113859.1 | - | 2 | 162 | 40030438 | ||
| BC114467.1 | - | 2 | 162 | 40030437 | ||
| CR457429.1 | - | 1 | 100 | |||
| X81422.1 | - | 2 | 163 | |||

PTPRCAP
Genomic Map