siRNA Details
| Gene Symbol: RARRES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002889.3 | NP_002880.1 | 3 | 330 | ||
| XM_017012491.1 | XP_016867980.1 | 3 | 341 | |||
| GenBank mRNA | AB015632.1 | - | 3 | 285 | ||
| AK092804.1 | - | 1 | 1497 | |||
| AK312197.1 | - | 3 | 284 | |||
| BC000069.2 | - | 3 | 261 | 3506067 | ||
| BM919890.1 | - | 2 | 265 | |||
| BX448753.2 | - | 3 | 330 | |||
| CR541992.1 | - | 2 | 213 | |||
| CR542026.1 | - | 2 | 213 | |||
| U77594.2 | - | 3 | 273 | |||

RARRES2
Genomic Map