siRNA Details
| Gene Symbol: RPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000999.3 | NP_000990.1 | 3 | 181 | ||
| NM_001035258.1 | NP_001030335.1 | 3 | 146 | |||
| GenBank mRNA | AK097905.1 | - | 2 | 346 | ||
| AK312119.1 | - | 3 | 149 | |||
| BC000603.2 | - | 3 | 118 | 3346447 | ||
| BQ276548.1 | - | 3 | 177 | |||
| BU532782.1 | - | 3 | 149 | |||
| CB124619.1 | - | 3 | 171 | |||
| CB128529.1 | - | 3 | 124 | |||
| Z26876.1 | - | 3 | 163 | |||

RPL38
Genomic Map