siRNA Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001022.3 | NP_001013.1 | 3 | 493 | ||
| NM_001321483.1 | NP_001308412.1 | 3 | 170 | |||
| NM_001321484.1 | NP_001308413.1 | 3 | 300 | |||
| NM_001321485.1 | NP_001308414.1 | 3 | 493 | |||
| XM_017027113.1 | XP_016882602.1 | 3 | 488 | |||
| GenBank mRNA | AA165492.1 | - | 1 | 47 | ||
| AK311786.1 | - | 3 | 156 | |||
| BC000023.2 | - | 3 | 190 | 3504083 | ||
| BC007615.2 | - | 2 | 121 | 3354280 | ||
| BC017386.1 | - | 3 | 159 | |||
| BC018616.1 | - | 3 | 493 | 4867036 | ||
| BG398397.1 | - | 3 | 300 | |||
| BM468237.1 | - | 3 | 264 | |||
| BP270469.1 | - | 3 | 156 | |||
| BU603942.1 | - | 3 | 159 | |||
| CB124389.1 | - | 3 | 199 | |||
| CB125125.1 | - | 3 | 170 | |||
| M81757.1 | - | 3 | 143 | |||

RPS19
Genomic Map