siRNA Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003083.3 | NP_003074.1 | 3, 4 | 427 | ||
| NR_030717.1 | 3, 4 | 324 | ||||
| GenBank mRNA | AK222926.1 | - | 3, 4 | 398 | ||
| AK314878.1 | - | 3, 4 | 412 | |||
| BC011868.2 | - | 3, 4 | 382 | 4545178 | ||
| BP234322.1 | - | 3, 4 | 323 | |||
| BX647416.1 | - | 3, 4 | 291 | |||
| CF454397.1 | - | 3, 4 | 427 | |||
| U44755.1 | - | 3, 4 | 411 | |||
| U44898.1 | - | 3, 4 | 393 | |||

SNAPC2
Genomic Map