siRNA Details
| Gene Symbol: SPRR2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024209.3 | NP_001019380.2 | 2 | 371 | ||
| GenBank mRNA | BC058018.1 | - | 2 | 386 | ||
| DA761491.1 | - | 2 | 370 | |||
| M20030.1 | - | 2 | 370 | |||
| Gene Symbol: SPRR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005988.2 | NP_005979.1 | 2 | 368 | ||

SPRR2E
Genomic Map