siRNA Details
| Gene Symbol: SPRR2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024209.3 | NP_001019380.2 | 2 | 515 | ||
| GenBank mRNA | BC058018.1 | - | 2 | 530 | ||
| M20030.1 | - | 2 | 514 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2E| Gene Symbol: SPRR2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024209.3 | NP_001019380.2 | 2 | 515 | ||
| GenBank mRNA | BC058018.1 | - | 2 | 530 | ||
| M20030.1 | - | 2 | 514 | |||