siRNA Details
| Gene Symbol: SULT1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001054.3 | NP_001045.1 | 5 | 485 | ||
| NM_177528.2 | NP_803564.1 | 5 | 758 | |||
| XM_017023600.1 | XP_016879089.1 | 5 | 494 | |||
| XM_017023601.1 | XP_016879090.1 | 5 | 922 | |||
| XM_017023602.1 | XP_016879091.1 | 6 | 624 | |||
| GenBank mRNA | BC113727.1 | - | 5 | 480 | 8322779 | |
| BC113729.1 | - | 5 | 480 | 8322781 | ||
| U28169.1 | - | 5 | 480 | |||
| U28170.1 | - | 5 | 468 | |||
| X78282.1 | - | 4 | 407 | |||

SULT1A2
Genomic Map