siRNA Details
| Gene Symbol: VPREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303509.1 | NP_001290438.1 | 2 | 358 | ||
| NM_007128.3 | NP_009059.1 | 2 | 372 | |||
| GenBank mRNA | BC017922.1 | - | 2 | 364 | ||
| CR456609.1 | - | 2 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPREB1| Gene Symbol: VPREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303509.1 | NP_001290438.1 | 2 | 358 | ||
| NM_007128.3 | NP_009059.1 | 2 | 372 | |||
| GenBank mRNA | BC017922.1 | - | 2 | 364 | ||
| CR456609.1 | - | 2 | 349 | |||