siRNA Details
| Gene Symbol: INSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005478.4 | NP_005469.2 | 2 | 386 | ||
| GenBank mRNA | AF133816.2 | - | 2 | 395 | ||
| AY359030.1 | - | 2 | 386 | |||
| BC101646.1 | - | 2 | 375 | 8069152 | ||
| BC101648.1 | - | 2 | 375 | 8069154 | ||

INSL5
Genomic Map