siRNA Details
| Gene Symbol: SLC30A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318949.1 | NP_001305878.1 | 9 | 1775 | ||
| NM_001318950.1 | NP_001305879.1 | 9 | 1920 | |||
| NM_001318951.1 | NP_001305880.1 | 8 | 1702 | |||
| NM_003459.4 | NP_003450.2 | 8 | 1838 | |||
| XM_005264547.1 | XP_005264604.1 | 7 | 1680 | |||
| XM_005264548.3 | XP_005264605.1 | 8 | 1666 | |||
| XM_006712100.2 | XP_006712163.1 | 7 | 3026 | |||
| XM_011533102.2 | XP_011531404.1 | 8 | 1835 | |||
| XM_011533103.2 | XP_011531405.1 | 7 | 1695 | |||
| XM_017004873.1 | XP_016860362.1 | 7 | 1561 | |||
| XM_017004874.1 | XP_016860363.1 | 8 | 1809 | |||
| XM_017004875.1 | XP_016860364.1 | 8 | 1593 | |||
| GenBank mRNA | AK127096.1 | - | 3 | 2467 | ||
| AK127300.1 | - | 4 | 2305 | |||
| BC028358.2 | - | 8 | 1826 | 4837595 | ||
| U76010.1 | - | 8 | 1737 | |||

SLC30A3
Genomic Map