siRNA Details
| Gene Symbol: HIST1H2AJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021066.2 | NP_066544.1 | 1 | 373 | ||
| GenBank mRNA | BC066232.1 | - | 1 | 373 | ||
| BC066233.1 | - | 1 | 373 | |||
| BC066234.1 | - | 1 | 373 | |||
| BC066235.1 | - | 1 | 373 | |||
| BC066236.1 | - | 1 | 373 | |||
| BC066237.1 | - | 1 | 373 | |||
| BC133048.1 | - | 1 | 462 | 40147070 | ||
| BC133050.1 | - | 1 | 462 | 40147072 | ||

HIST1H2AJ
Genomic Map