siRNA Details
| Gene Symbol: HIST1H2AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003513.2 | NP_003504.2 | 1 | 168 | ||
| GenBank mRNA | AK311785.1 | - | 1 | 219 | ||
| BC125140.1 | - | 1 | 172 | 40119630 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AB| Gene Symbol: HIST1H2AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003513.2 | NP_003504.2 | 1 | 168 | ||
| GenBank mRNA | AK311785.1 | - | 1 | 219 | ||
| BC125140.1 | - | 1 | 172 | 40119630 | ||