siRNA Details
| Gene Symbol: HIST1H2AM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003514.2 | NP_003505.1 | 1 | 419 | ||
| GenBank mRNA | AK312224.1 | - | 1 | 419 | ||
| BC032686.1 | - | 1 | 434 | |||
| BC032756.1 | - | 1 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AM| Gene Symbol: HIST1H2AM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003514.2 | NP_003505.1 | 1 | 419 | ||
| GenBank mRNA | AK312224.1 | - | 1 | 419 | ||
| BC032686.1 | - | 1 | 434 | |||
| BC032756.1 | - | 1 | 434 | |||