siRNA Details
Gene Symbol: HIST1H2BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003519.3 | NP_003510.1 | 1 | 404 | ||
GenBank mRNA | BC093759.1 | - | 1 | 461 | ||
BC093761.1 | - | 1 | 460 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST1H2BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003519.3 | NP_003510.1 | 1 | 404 | ||
GenBank mRNA | BC093759.1 | - | 1 | 461 | ||
BC093761.1 | - | 1 | 460 |