siRNA Details
| Gene Symbol: HIST1H2BI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003525.2 | NP_003516.1 | 1 | 251 | ||
| GenBank mRNA | BC101653.1 | - | 1 | 311 | 8069159 | |
| BC101655.1 | - | 1 | 311 | 8069161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2BI| Gene Symbol: HIST1H2BI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003525.2 | NP_003516.1 | 1 | 251 | ||
| GenBank mRNA | BC101653.1 | - | 1 | 311 | 8069159 | |
| BC101655.1 | - | 1 | 311 | 8069161 | ||