siRNA Details
| Gene Symbol: HIST1H4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003538.3 | NP_003529.1 | 1 | 321 | ||
| GenBank mRNA | BC066248.1 | - | 1 | 321 | 7002088 | |
| BC066249.1 | - | 1 | 321 | 7002089 | ||
| BC067495.1 | - | 1 | 321 | 7002090 | ||
| BC069654.1 | - | 1 | 321 | 7262752 | ||
 Back To Top
 Back To Top
 HIST1H4A
HIST1H4A Genomic Map
Genomic Map Back To Top
 Back To Top