siRNA Details
| Gene Symbol: PGLYRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005091.2 | NP_005082.1 | 1 | 515 | ||
| GenBank mRNA | AF076483.1 | - | 1 | 282 | ||
| AF242517.1 | - | 1 | 267 | |||
| AY358936.1 | - | 1 | 263 | |||
| BC096154.1 | - | 1 | 275 | 40003108 | ||
| BC096155.3 | - | 1 | 275 | 40003109 | ||
| BC096156.1 | - | 1 | 275 | 40003112 | ||
| BC096157.1 | - | 1 | 275 | 40003114 | ||
| BC101845.1 | - | 1 | 277 | 8069351 | ||
| BC101847.1 | - | 1 | 277 | 8069353 | ||
| DN998360.1 | - | 1 | 551 | |||

PGLYRP1
Genomic Map