siRNA Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258450.1 | NP_001245379.1 | 4, 5 | 1167 | ||
| NM_001258451.1 | NP_001245380.1 | 4, 5 | 1238 | |||
| NM_004203.4 | NP_004194.3 | 4, 5 | 1374 | |||
| NM_182687.2 | NP_872629.1 | 4, 5 | 1374 | |||
| XM_011522734.2 | XP_011521036.1 | 4, 5 | 1335 | |||
| XM_011522735.2 | XP_011521037.1 | 4, 5 | 1161 | |||
| XM_011522736.2 | XP_011521038.1 | 3, 4 | 1168 | |||
| GenBank mRNA | AK097642.1 | - | 4, 5 | 1123 | ||
| AK098452.1 | - | 4, 5 | 1226 | |||
| AK294379.1 | - | 3, 4 | 1135 | |||
| AK301926.1 | - | 4, 5 | 1043 | |||
| AK303005.1 | - | 4, 5 | 1252 | |||
| AK309964.1 | - | 3, 4 | 1168 | |||
| BC121161.1 | - | 3, 4 | 1062 | |||
| BC121162.1 | - | 3, 4 | 1062 | |||
| BC130011.1 | - | 3, 4 | 1062 | |||
| BC153823.1 | - | 3, 4 | 1062 | |||
| U56816.1 | - | 3, 4 | 1078 | |||

PKMYT1
Genomic Map