siRNA Details
| Gene Symbol: ISG15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005101.3 | NP_005092.1 | 2 | 194 | ||
| GenBank mRNA | AY168648.1 | - | 1 | 87 | ||
| BC009507.2 | - | 2 | 128 | 3545944 | ||
| BM712238.1 | - | 2 | 195 | |||
| BT007297.1 | - | 1 | 87 | |||
| BU161226.1 | - | 2 | 151 | |||
| M13755.1 | - | 2 | 162 | |||

ISG15
Genomic Map