siRNA Details
| Gene Symbol: CHAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005483.2 | NP_005474.2 | 3 | 374 | ||
| XM_011527605.2 | XP_011525907.1 | 3 | 441 | |||
| XM_011527606.2 | XP_011525908.1 | 3 | 441 | |||
| XM_011527607.2 | XP_011525909.1 | 3 | 441 | |||
| GenBank mRNA | BC041606.1 | - | 3 | 299 | ||
| BC052620.1 | - | 3 | 373 | |||
| BC067093.1 | - | 3 | 375 | 6339027 | ||
| BQ889365.1 | - | 3 | 374 | |||
| U20979.1 | - | 3 | 260 | |||

CHAF1A
Genomic Map