siRNA Details
| Gene Symbol: IFITM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021034.2 | NP_066362.2 | 2 | 500 | ||
| NR_049759.1 | 2 | 495 | ||||
| GenBank mRNA | AK292173.1 | - | 2 | 444 | ||
| BC006794.1 | - | 2 | 458 | 2986145 | ||
| BC008417.1 | - | 2 | 443 | 4075453 | ||
| BC022439.1 | - | 2 | 449 | 4282809 | ||
| BC070243.1 | - | 2 | 434 | 30401041 | ||
| BQ937265.1 | - | 2 | 303 | |||
| DB261103.1 | - | 2 | 500 | |||
| X57352.1 | - | 2 | 636 | |||

IFITM3
Genomic Map