siRNA Details
| Gene Symbol: CCDC85B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006848.2 | NP_006839.2 | 1 | 988 | ||
| GenBank mRNA | BC008796.2 | - | 1 | 753 | 3617697 | |
| U63825.1 | - | 1 | 636 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC85B| Gene Symbol: CCDC85B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006848.2 | NP_006839.2 | 1 | 988 | ||
| GenBank mRNA | BC008796.2 | - | 1 | 753 | 3617697 | |
| U63825.1 | - | 1 | 636 | |||