siRNA Details
| Gene Symbol: PPP1R14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138689.2 | NP_619634.1 | 1 | 348 | ||
| GenBank mRNA | BC014522.2 | - | 1 | 200 | 3831227 | |
| BC071884.1 | - | 1 | 103 | |||
| BC094817.1 | - | 1 | 231 | 5466876 | ||
| BM847221.1 | - | 1 | 348 | |||
| BT009914.1 | - | 1 | 80 | |||
| X91195.1 | - | 1 | 332 | |||

PPP1R14B
Genomic Map