siRNA Details
| Gene Symbol: DUX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012149.2 | NP_036281.2 | 1 | 220 | ||
| Gene Symbol: DUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012146.1 | NP_036278.1 | 1 | 251 | ||
| GenBank mRNA | AJ001481.1 | - | 1 | 251 | ||
| Gene Symbol: DUX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012148.2 | NP_036280.2 | 1 | 220 | ||

DUX5
Genomic Map