siRNA Details
| Gene Symbol: OR7E24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079935.1 | NP_001073404.1 | 1 | 55 | ||
| GenBank mRNA | BC146920.1 | - | 1 | 169 | 9056848 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR7E24| Gene Symbol: OR7E24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079935.1 | NP_001073404.1 | 1 | 55 | ||
| GenBank mRNA | BC146920.1 | - | 1 | 169 | 9056848 | |