siRNA Details
Gene Symbol: IL17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013278.3 | NP_037410.1 | 3 | 859 | ||
GenBank mRNA | AF152099.1 | - | 3 | 859 | ||
AY358471.1 | - | 3 | 859 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IL17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013278.3 | NP_037410.1 | 3 | 859 | ||
GenBank mRNA | AF152099.1 | - | 3 | 859 | ||
AY358471.1 | - | 3 | 859 |