siRNA Details
| Gene Symbol: GPR78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080819.4 | NP_543009.2 | 3 | 1544 | ||
| NR_045511.2 | 3 | 851 | ||||
| GenBank mRNA | AF411107.1 | - | 3 | 1051 | ||
| AK128807.1 | - | 3 | 1544 | |||
| AK307281.1 | - | 3 | 851 | |||
| AK308759.1 | - | 2 | 529 | |||
| AK313038.1 | - | 3 | 1119 | |||
| AY359107.1 | - | 3 | 1345 | |||
| BC057778.1 | - | 3 | 1468 | 30343110 | ||
| BC069343.1 | - | 3 | 1051 | 7262307 | ||
| BC069813.1 | - | 3 | 1051 | 7262319 | ||
| JF810885.1 | - | 3 | 1051 | |||
More Information
Set Membership: |
|

GPR78
Genomic Map