siRNA Details
| Gene Symbol: GPR78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080819.4 | NP_543009.2 | 3 | 1928 | ||
| NR_045511.2 | 3 | 1235 | ||||
| GenBank mRNA | AK128807.1 | - | 3 | 1928 | ||
| AK308759.1 | - | 2 | 913 | |||
| BC057778.1 | - | 3 | 1852 | 30343110 | ||
More Information
Set Membership: |
|

GPR78
Genomic Map