siRNA Details
| Gene Symbol: DHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014475.3 | NP_055290.1 | 2 | 174 | ||
| XM_005258748.4 | XP_005258805.1 | 2 | 735 | |||
| XM_017026598.1 | XP_016882087.1 | 2 | 734 | |||
| GenBank mRNA | AB021933.1 | - | 2 | 134 | ||
| BC032730.1 | - | 2 | 185 | 5530225 | ||

DHDH
Genomic Map